Learn More
Guanethidine sulfate, Thermo Scientific™
Specifications
Specifications
| Packaging | Glass Bottle |
| Certificate of Analysis | Available |
| Concentration or Composition Notes | Titration with HClO4 |
| Physical Form | Crystalline Powder/Crystals |
| Concentration | 97.5% to 102.5% |
| Color | White to Light Beige |
| Quantity | 1 g |
| Formula Weight | 296.39 |
| Chemical Name or Material | Guanethidine sulfate |
Safety and Handling
| Hazard Category | GHS07: Exclamation mark |
| ShelfLife | 2 years |
| EINECSNumber | 211-442-0 |
| RTECSNumber | MF3150000 |
| Recommended Storage | Normal conditions |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.