Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Guanfacine hydrochloride
GREENER_CHOICESDP

Catalog No. 103050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-305-0 50 mg
10-301-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-305-0 Supplier R&D Systems™ Supplier No. 1030/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α2A agonist

Guanfacine hydrochloride is a selective α2A-adrenoceptor agonist (Kd = 31 nM). Displays 60-fold selectivity over α2B-adrenoceptors.

Chemical Identifiers

CAS 29110-48-3
Molecular Formula C9H10Cl3N3O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 282.56
Purity >99%
Molecular Weight (g/mol) 282.549
Chemical Name or Material Guanfacine hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.