Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Carbonate, 99+%

Catalog No. AC120220051
Click to view available options
Quantity:
250 g
1 kg
5 kg

CAS: 593-85-1 | 0·5 H2CO3 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-85-1
Molecular Formula 0·5 H2CO3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00013029

Specifications

Melting Point 267.0°C to 270.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic drum
Linear Formula [C(NH2)3]2CO3
Quantity 5 kg
Beilstein 03,82
Solubility Information Solubility in water: 450g/L (20 c). Other solubilities: slightly soluble in alcohol and acetone
Formula Weight 90.08
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Guanidine carbonate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-813-7
RTECSNumber FG1750000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.