Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2)

Catalog No. p-4509622
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum

Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Quantity: 1kg, Packaging: Plastic bottle, White to Yellow, Melting Point: 179.0 deg.C to 186.0 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Assay Percent Range: 0.5% max. Ammonium chloride (Argentometry) | CAS: 50-01-1 | CH5N3·HCl | 95.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH5N3·HCl
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 179.0°C to 186.0°C
Color White to Yellow
Density 1.3400g/mL
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. Ammonium chloride (Argentometry)
Packaging Plastic bottle
Linear Formula H2NC(=NH)NH2·HCl
Quantity 250 g
Beilstein 03,82
Specific Gravity 1.34
Merck Index 15,4597
Solubility Information Solubility in water: 2280g/L (20°C). Other solubilities: 300g/L ethanol (30°C), 19.5g/100g dimethylformamide (30°C), <0.05g/100g acetone -<0.05g/100g benzene (30°C), insoluble in trichloroethylene
Formula Weight 95.53
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Guanidine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
EINECSNumber 200-002-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.