Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Hydrochloride, >99.5%, Molecular Biology Grade

Catalog No. aaj7582322
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Poly pail
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 50-01-1 | CH6ClN3 | 95.53 g/mol

  • Used for extraction of RNA.
  • Preparation of high molecular weight genomic DNA.
  • Solubilizing inclusion bodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used for extraction of RNA
  • Preparation of high molecular weight genomic DNA
  • Solubilizing inclusion bodies
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 185°C to 188°C
Density 1.344
Packaging Plastic bottle
Linear Formula (NH2)2C=NH·HCl
Quantity 100 g
Beilstein 3591990
Formula Weight 95.53
Percent Purity ≥99.5%
Grade Molecular Biology
Physical Form Crystals
Chemical Name or Material Guanidine Hydrochloride
Show More Show Less
Hazard Category H302+H332-H315-H319-H500
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-002-3
RTECSNumber MF4300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.