Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™

Catalog No. BP1781 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP178-1 1 kg
BP178-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP178-1
Add to Cart

For isolation of RNA from cell extracts | CAS: 50-01-1 | CH6ClN3

For isolation of RNA from cell extracts

Guanidine Hydrochloride is a strong protein-denaturing agent used in the isolation of RNA from cell extracts. RNase activity is inhibited in the presence of this reagent.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guanidine Hydrochloride
CAS 50-01-1
Assay Percent Range ≥99.5%
Synonym guanidine hydrochloride
Quantity 1 kg
Merck Index 15, 4597
Melting Point 178°C
Molecular Formula CH6ClN3
Formula Weight 95.53
Physical Form Solid
Color White
Purity ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.