Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Thiocyanate, Molecular Biology Grade, PROMO, Thermo Scientific Chemicals

Catalog No. J6510418PM
Click to view available options
Quantity:
50 g

A protein denaturant

Applications

Guanidine thiocyanate is used for RNA isolation and can solubilize proteins. When the compound is used to solubilize proteins it can be removed by dialysis against urea without difficulty. The hybridization of guanidine thiocyanate-solubilize hepatocytes can be used as a method to quantify mRNA from hepatocytes. Blood sample lysate stored in guanidine thiocyanate has been shown to maintain nucleic acid integrity. Research describes guanidine thiocyanate to be ineffective for RNA isolation of plant tissue with a high levels of secondary metabolites and storage proteins.

Solubility

Soluble in water (1420 g/L) at 20°C, and ethanol (5%).

Notes

Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-84-0
Molecular Formula CH5N3·CHNS
MDL Number MFCD00013027
Synonym Guanidine rhodanide

Specifications

Melting Point 118°C to 122°C
Color White
Quantity 50 g
Beilstein 3563461
Sensitivity Light Sensitive
Solubility Information Soluble in water at 20°C, and ethanol.
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material Guanidine thiocyanate
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 209-812-1
RTECSNumber XL1225000
TSCA Yes

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.