Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 26045 |
| Species | Human |
| Purity | Immunogen affinity purified |
| Common Name | LRRTM2 |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP2-41129. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | leucine-rich repeat transmembrane neuronal protein 2, leucine rich repeat transmembrane neuronal 2 |
| Gene Symbol | LRRTM2 |
| Quantity | 50 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
