Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanine 98.0+%, TCI America™
SDP

Catalog No. G0169250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0169250G 250 g
G01691G 1 g
G016925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0169250G Supplier TCI America Supplier No. G0169250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73-40-5
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00071533
InChI Key UYTPUPDQBNUYGX-UHFFFAOYSA-N
Synonym guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence
PubChem CID 764
ChEBI CHEBI:16235
IUPAC Name 2-amino-6,7-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Quantity 250 g
Formula Weight 151.13
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Guanine
EINECSNumber (5)-1136&(9)-0328
RTECSNumber MF8260000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.