Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanosine-5'-DiPhosphate Disodium Salt, PROMO, Thermo Scientific Chemicals

Catalog No. J61646MEPM
Click to view available options
Quantity:
100 mg
500 mg

Applications

Guanosine-5'-diphosphate disodium salt is used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors. It is also used to study cell signaling processes mediate by guanine nucleotide exchange factors.

Solubility

Soluble in water

Notes

Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7415-69-2
Molecular Formula C10H13N5Na2O11P2
Molecular Weight (g/mol) 487.17 g/mol
MDL Number MFCD00066538
InChI Key LTZCGDIGAHOTKN-KHRSEZDTNA-L
Synonym 5-gdp pound not2na
PubChem CID 134129638
IUPAC Name disodium;(2R,3S,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Color White
Quantity 500 mg
Solubility Information Soluble in water
Physical Form Powder
Chemical Name or Material Guanosine-5'-diphosphate disodium salt
EINECSNumber 231-026-2
TSCA No
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.