Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylamine N-oxide dihydrate, 98%

Catalog No. AC421891000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

Catalog No. AC421891000

Supplier: Thermo Scientific Chemicals 421891000

Only null left
Add to Cart

CAS: 62637-93-8 | C3H13NO3 | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62637-93-8
Molecular Formula C3H13NO3
Molecular Weight (g/mol) 111.14
MDL Number MFCD00149077
InChI Key PGFPZGKEDZGJQZ-UHFFFAOYSA-N
Synonym trimethylamine-n-oxide dihydrate, trimethylamine n-oxide dihydrate, trimethylamine oxide dihydrate, trimethylamine oxide, dihydrate, n,n-dimethylmethanamine oxide, dihydrate, unii-c73wz0186w, trimethylammoniumoxid hydrat czech, methanamine oxide, n,n-dimethyl, dihydrate, methanamine, n,n-dimethyl-, n-oxide, dihydrate, n,n-dimethylmethanamine oxide dihydrate
PubChem CID 198430
SMILES O.O.C[N+](C)(C)[O-]

Specifications

Melting Point 94.0°C to 98.0°C
Color White
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (Total base)
Packaging Glass bottle
Linear Formula (CH3)3NO·2H2O
Quantity 100 g
Fieser 01,1230; 02,434; 03,309; 06,624; 07,392; 08,507; 09,488; 14,329
Merck Index 15,9881
IUPAC Name N,N-dimethylmethanamine oxide;dihydrate
Formula Weight 111.14
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Trimethylamine N-oxide dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.