Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanylate Cyclase 2E Mouse anti-Mouse, Bovine, Human, Unconjugated, Clone: 5H2, MilliporeSigma™

Catalog No. MABN243625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2436-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2436-25 Supplier MilliporeSigma Supplier No. MABN243625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Peripherin-2, clone 5H2, Cat. No. MABN2435, is a highly specific mouse monoclonal antibody that targets Peripherin-2 and has been tested in Immunofluorescence, Radioimmunoassay, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Guanylate Cyclase 2E
Applications Immunofluorescence, Immunoradiometric Assay, Western Blot
Classification Monoclonal
Clone 5H2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias RETGC-1;Guanylate cyclase-E;Guanylate cyclase-2E;GC-E
Gene Symbols GUCY2D
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 48 amino acids from the C-terminal end of human Guanylate cyclase-1.
Molecular Weight of Antigen ∽39 kDa observed; 38.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 8A5 detects Guanylate cyclase-1 in Bovine, Human, and murine species. It targets an epitope within 48 amino acids from the C-terminal end.
Target Species Mouse, Bovine, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.