Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylate kinase Antibody (OTI4A8), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270869MFV610
Description
Guanylate kinase Monoclonal antibody specifically detects Guanylate kinase in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Guanylate kinase | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI4A8 | |
Flow Cytometry | |
EC 2.7.4.8, FLJ42686, FLJ43710, GMK, GMP kinase, guanylate kinase, guanylate kinase 1 | |
Full length human recombinant protein of human GUK1 (NP_000849) produced in HEK293T cell. | |
0.1 mL | |
Cancer, Signal Transduction | |
2987 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction