Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylate kinase Antibody (OTI4A8), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270869PECY7
Description
Guanylate kinase Monoclonal specifically detects Guanylate kinase in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Guanylate kinase | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
GUK1 | |
Full length recombinant protein of human Guanylate kinase (NP_000849) produced in HEK293T cells. | |
0.1 mL | |
2987.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI4A8 | |
Flow Cytometry | |
EC 2.7.4.8, FLJ42686, FLJ43710, GMK, GMP kinase, guanylate kinase, guanylate kinase 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction