Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylin Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Guanylin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Guanylin Polyclonal specifically detects Guanylin in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Guanylin | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
2980 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
GCAP-I, Guanylate cyclase activator 2A, guanylate cyclase activator 2A (guanylin), Guanylate cyclase-activating protein 1, Guanylate cyclase-activating protein I, guanylin, GUCA2guanylate cyclase activator 2A (guanylin 2, intestinal, heat-stable), STARA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 22-115 of human Guanylin (NP_291031.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title