Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanylin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981524
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1524 Supplier Bioss Supplier No. BS11621R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Endogenous activator of intestinal guanylate cyclase. It stimulates this enzyme through the same receptor binding region as the heat-stable enterotoxins.

Specifications

Antigen Guanylin
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GUCA2A
Gene Accession No. Q02747
Gene Alias GCAP-I; GUANYL; guanylate cyclase activator 2 (guanylin 2, intestinal, heatstable); guanylate cyclase activator 2 (guanylin); Guanylate cyclase activator 2A; guanylate cyclase activator 2A (guanylin 2, intestinal, heat-stable); guanylate cyclase activator 2A (guanylin); Guanylate cyclase-activating protein 1; Guanylate cyclase-activating protein I; Guanylin; Guca2; Guca2a; HMW-guanylin; prepro-guanylin; STARA
Gene Symbols GUCA2A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Guanylin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2980
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.