Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylyl Cyclase alpha 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP26874225UL
Description
Guanylyl Cyclase alpha 1 Polyclonal antibody specifically detects Guanylyl Cyclase alpha 1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Guanylyl Cyclase alpha 1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
EC 4.6.1.2, GC-S-alpha-1, GCS-alpha-1, GCS-alpha-3, guanylate cyclase 1, soluble, alpha 3, guanylate cyclase soluble subunit alpha-3, GUC1A3GUCA3, GUCSA3GC-SA3, GUCY1A1, Soluble guanylate cyclase large subunit | |
This antibody was developed against a recombinant protein corresponding to amino acids: PPCFHNDCSEFVNQPYLLYSVHMKSTKPSLSPSKPQSSLVIPTSLFCKTFPFHFMFDKDMTILQFGNGIR | |
25 μL | |
Signal Transduction, Vision | |
2982 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction