Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylyl Cyclase alpha 1 Antibody (SR2180), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Guanylyl Cyclase alpha 1 |
---|---|
Clone | SR2180 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Guanylyl Cyclase alpha 1 Monoclonal antibody specifically detects Guanylyl Cyclase alpha 1 in Human samples. It is validated for Western BlotSpecifications
Guanylyl Cyclase alpha 1 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 4.6.1.2, GC-S-alpha-1, GCS-alpha-1, GCS-alpha-3, guanylate cyclase 1, soluble, alpha 3, guanylate cyclase soluble subunit alpha-3, GUC1A3GUCA3, GUCSA3GC-SA3, GUCY1A1, Soluble guanylate cyclase large subunit | |
A synthesized peptide derived from human Guanylyl Cyclase alpha 1 (Uniprot #: Q02108) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2180 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction, Vision | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2982 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title