Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanylyl Cyclase alpha 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982961
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2961 Supplier Bioss Supplier No. BS13545R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha and a beta subunit, catalyzes the conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide and nitrovasodilator drugs.

Specifications

Antigen Guanylyl Cyclase alpha 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GUCY1A2
Gene Accession No. P33402
Gene Alias 6330407I18Rik; A230060L24Rik; BOS_14298; GC-SA2; GCS-alpha-2; guanylate cyclase 1 soluble subunit alpha 2; guanylate cyclase 1, soluble, alpha 2; Guanylate cyclase soluble subunit alpha-2; GUC1A2; GUCSA2; Gucy1a2; LOC100357621; soluble guanylyl cyclase alpha2 subunit
Gene Symbols GUCY1A2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Guanylyl Cyclase alpha 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2977
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.