Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylyl Cyclase beta 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318694
This item is not returnable.
View return policy
Description
Guanylyl Cyclase beta 1 Polyclonal antibody specifically detects Guanylyl Cyclase beta 1 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Dot BlotSpecifications
Guanylyl Cyclase beta 1 | |
Polyclonal | |
Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:200, Dot Blot 1:10000 | |
EC 4.6.1.2, GC-SB3, GCS-beta-1, GCS-beta-3, guanylate cyclase 1, soluble, beta 3, guanylate cyclase soluble subunit beta-1, Guanylate cyclase soluble subunit beta-3, GUC1B3GUCB3, GUCSB3GC-S-beta-1, GUCY1B1, Soluble guanylate cyclase small subunit | |
Synthetic peptide taken within amino acid region 175-225 on rat guanylate cyclase soluble subunit beta-1 protein. | |
100 μg | |
Signal Transduction | |
2983 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Dot Blot | |
Unconjugated | |
Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary). | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction