Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylyl Cyclase beta 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP158869
Description
Guanylyl Cyclase beta 1 Polyclonal specifically detects Guanylyl Cyclase beta 1 in Human, Mouse samples. It is validated for Western Blot.Specifications
Guanylyl Cyclase beta 1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 4.6.1.2, GC-SB3, GCS-beta-1, GCS-beta-3, guanylate cyclase 1, soluble, beta 3, guanylate cyclase soluble subunit beta-1, Guanylate cyclase soluble subunit beta-3, GUC1B3GUCB3, GUCSB3GC-S-beta-1, GUCY1B1, Soluble guanylate cyclase small subunit | |
Rabbit | |
71 kDa | |
100 μL | |
Primary | |
This product is specific to Subunit or Isoform: beta-1. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q02153 | |
GUCY1B3 | |
Synthetic peptides corresponding to GUCY1B3(guanylate cyclase 1, soluble, beta 3) The peptide sequence was selected from the N terminal of GUCY1B3 (NP_000848). Peptide sequence LIEEKESKEEDFYEDLDRFEENGTQESRISPYTFCKAFPFHIIFDRDLVV. | |
Affinity purified | |
RUO | |
2983 | |
Centrifuge prior to opening. Reconstitute with sterilized PBS to a final concentration of 1mg/ml. Vortex followed by Centrifuge again to pellet the solution. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction