Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylyl Cyclase beta 1 Antibody (SR2169), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32198750UL
Description
Guanylyl Cyclase beta 1 Monoclonal antibody specifically detects Guanylyl Cyclase beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Guanylyl Cyclase beta 1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR2169 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
EC 4.6.1.2, GC-SB3, GCS-beta-1, GCS-beta-3, guanylate cyclase 1, soluble, beta 3, guanylate cyclase soluble subunit beta-1, Guanylate cyclase soluble subunit beta-3, GUC1B3GUCB3, GUCSB3GC-S-beta-1, GUCY1B1, Soluble guanylate cyclase small subunit | |
A synthesized peptide derived from human Guanylyl Cyclase beta 1 (Uniprot #: Q02153) | |
50 μg | |
Signal Transduction | |
2983 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction