Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylyl Cyclase beta 1 Rabbit anti-Human, Mouse, Rat, Clone: 5O8D9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Guanylyl Cyclase beta 1 |
---|---|
Clone | 5O8D9 |
Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Guanylyl Cyclase beta 1 Monoclonal antibody specifically detects Guanylyl Cyclase beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Guanylyl Cyclase beta 1 | |
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 4.6.1.2, GC-SB3, GCS-beta-1, GCS-beta-3, guanylate cyclase 1, soluble, beta 3, guanylate cyclase soluble subunit beta-1, Guanylate cyclase soluble subunit beta-3, GUC1B3GUCB3, GUCSB3GC-S-beta-1, GUCY1B1, Soluble guanylate cyclase small subunit | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Guanylyl Cyclase beta 1 (Q02153). MYGFVNHALELLVIRNYGPEVWEDIKKEAQLDEEGQFLVRIIYDDSKTYDLVAAASKVLNLNAGEILQMFGKMFFVFCQESGYDTILRVLGSNVREFLQN | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
5O8D9 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2983 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title