Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guar Gum, NF, Spectrum™ Chemical
SDP

Catalog No. 18603087
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail

G1044, 9000-30-0

Spectrum™ Chemical Guar Gum, NF is a binder or disintegrator in solid dosage pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-30-0
Molecular Formula C10H14N5Na2O12P3
Molecular Weight (g/mol) 535.15
InChI Key JEKDCIBJADJZSK-UHFFFAOYNA-L
IUPAC Name disodium hydrogen [({[5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
SMILES [Na+].[Na+].NC1=C2N=CN(C3CC(O)C(COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C2=NC=N1

Specifications

CAS Max % 100%
Quantity 2.5 kg
Grade NF
Loss on Drying 15%
Packaging Poly Pail
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.