Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Guardian™ 2000 Hotplate

Click to view available options
Temperature (Metric):
70°C to 500°C
Size:
4 x 4 in.
7 x 7 in.
Description
- Applications: Boiling Liquids, Basic Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
- Display: Prominent hot top indicator light, green indicator lights when unit is heating and stirring
- Operation: Simple knob control, stirring up to 1600 rpm, temperature up to 500°C (ceramic top) or 380°C (ceramic coated stainless steel top), power cord (included), stirring models include PTFE coated stir bar
- Communication: N/A
- Construction: Durable metal housing, PET front panel, control knobs, ON/OFF switch for heater disconnect, built in support rod holder, easy to clean resin, ceramic or ceramic coated stainless steel tops
- Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring

Specifications
Specifications
Top Plate Material | Ceramic |
Temperature (English) | 158°F to 932°F |
Controller Type | Analog |
Size | 4 x 4 in. |
Temperature (Metric) | 70°C to 500°C |
Weight (English) | 5.5 lb. |
Weight (Metric) | 2.5 kg |
Wattage | 505 W |
Amperage | 5 A |
Frequency | 50/60 Hz |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction