Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Guardian™ 2000 Hotplate and Stirrer
Shop All OHAUS Products

Click to view available options
Temperature (Metric):
50°C to 380°C
70°C to 500°C
Size:
135 mm dia.
4 x 4 in.
7 x 7 in.
Description
- Applications: Boiling Liquids, Basic Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
- Display: Prominent hot top indicator light, green indicator lights when unit is heating and stirring
- Operation: Simple knob control, stirring up to 1600 rpm, temperature up to 500°C (ceramic top) or 380°C (ceramic coated stainless steel top), power cord (included), stirring models include PTFE coated stir bar
- Communication: N/A
- Construction: Durable metal housing, PET front panel, control knobs, ON/OFF switch for heater disconnect, built in support rod holder, easy to clean resin, ceramic or ceramic coated stainless steel tops
- Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring
Specifications
Specifications
| Top Plate Material | Ceramic-coated Stainless Steel |
| Voltage | 115 V |
| Controller Type | Analog |
| Model | e-G21HSRDS |
| Size | 135 mm dia. |
| Temperature (Metric) | 50°C to 380°C |
| Temperature (English) | 122°F to 716°F |
| Frequency | 60 Hz |
| Wattage | 535 W |
| Weight (English) | 5.5 lb. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction