Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guardian Protective Wear Hoods
Click to view available options
Color:
Lime Yellow
Olive Drab
Yellow
Includes:
Drawstring
Waterproof seal, snap for attaching to collar
Specifications
Specifications
| Size | Universal |
| Color | Lime Yellow |
| Seam Type | Sewn and taped |
| Quantity | 1 Ea. |
| Includes | Waterproof seal, snap for attaching to collar |
| For Use With (Application) | Wet environment |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction