Learn More
Invitrogen™ GUCA2A Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Specifications
Specifications
| Antigen | GUCA2A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GUCA2A |
| Gene Accession No. | P28902, P33680, Q02747 |
| Gene Alias | GCAP-I; GUANYL; guanylate cyclase activator 2 (guanylin 2, intestinal, heatstable); guanylate cyclase activator 2 (guanylin); guanylate cyclase activator 2A; guanylate cyclase activator 2A (guanylin 2, intestinal, heat-stable); guanylate cyclase activator 2a (guanylin); guanylate cyclase-activating protein 1; Guanylate cyclase-activating protein I; guanylin; GUCA2; Guca2a; HMW-guanylin; prepro-guanylin; STARA |
| Gene Symbols | GUCA2A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.