Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GUCA2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107138
Description
Sequence homology: Cow: 83%; Guinea Pig: 77%; Horse: 85%; Human: 100%; Mouse: 91%; Pig: 82%; Rabbit: 83%; Rat: 100%; Sheep: 92%; Yeast: 90%.
Endogenous activator of intestinal guanylate cyclase. It stimulates this enzyme through the same receptor binding region as the heat-stable enterotoxins.
Specifications
| GUCA2A | |
| Polyclonal | |
| Unconjugated | |
| GUCA2A | |
| GCAP-I; GUANYL; guanylate cyclase activator 2 (guanylin 2, intestinal, heatstable); guanylate cyclase activator 2 (guanylin); guanylate cyclase activator 2A; guanylate cyclase activator 2A (guanylin 2, intestinal, heat-stable); guanylate cyclase activator 2a (guanylin); guanylate cyclase-activating protein 1; Guanylate cyclase-activating protein I; guanylin; GUCA2; Guca2a; HMW-guanylin; prepro-guanylin; STARA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2980 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q02747 | |
| GUCA2A | |
| Synthetic peptide located within the following region: FAPIPGEPVV PILCSNPNFP EELKPLCKEP NAQEILQRLE EIAEDPGTCE. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction