Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GUCA2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103882 Supplier Invitrogen™ Supplier No. PA5103882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GUCA2B.

Endogenous activator of intestinal guanylate cyclase. It stimulates this enzyme through the same receptor binding region as the heat-stable enterotoxins. May be a potent physiological regulator of intestinal fluid and electrolyte transport. May be an autocrine/paracrine regulator of intestinal salt and water transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUCA2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GUCA2B
Gene Accession No. O09051, P70668, Q16661
Gene Alias AV066530; Gcap2; GCAP-II; granule cell antiserum positive 2; guanylate cyclase activator 2B; guanylate cyclase activator 2b (retina); guanylate cyclase activator 2B (uroguanylin); Guanylate cyclase C-activating peptide 2; Guanylate cyclase C-activating peptide II; Guca2b; prepro-uroguanylin; UGN; Uroguanylin
Gene Symbols GUCA2B
Host Species Rabbit
Immunogen A synthesized peptide derived from human GUCA2B(Accession Q16661), corresponding to amino acid residues V15-A65.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14916, 2981, 64055
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.