Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GUCA2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568208 Supplier Invitrogen™ Supplier No. PA568208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GUCA2B Polyclonal Antibody for Western Blot

Endogenous activator of intestinal guanylate cyclase. It stimulates this enzyme through the same receptor binding region as the heat-stable enterotoxins. May be a potent physiological regulator of intestinal fluid and electrolyte transport. May be an autocrine/paracrine regulator of intestinal salt and water transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUCA2B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GUCA2B
Gene Accession No. Q16661
Gene Alias AV066530; Gcap2; GCAP-II; granule cell antiserum positive 2; guanylate cyclase activator 2B; guanylate cyclase activator 2b (retina); guanylate cyclase activator 2B (uroguanylin); Guanylate cyclase C-activating peptide 2; Guanylate cyclase C-activating peptide II; Guca2b; prepro-uroguanylin; UGN; Uroguanylin
Gene Symbols GUCA2B
Host Species Rabbit
Immunogen Synthesized peptide derived from human GUCA2B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2981
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.