Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GUCY1A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546474 Supplier Invitrogen™ Supplier No. PA546474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KNFHNISNRC SYADHSNKEE IEDVSGILQC TANILGLKFE EIQKRFGEEF Sequence homology: Cow: 86%; Dog: 85%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 100%.

Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha and a beta subunit, catalyzes the conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide and nitrovasodilator drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUCY1A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GUCY1A2
Gene Accession No. P33402
Gene Alias 6330407I18Rik; A230060L24Rik; GC-SA2; GCS-alpha-2; guanylate cyclase 1 soluble subunit alpha 2; guanylate cyclase 1, soluble, alpha 2; guanylate cyclase soluble subunit alpha-2; GUC1A2; GUCSA2; Gucy1a2; soluble guanylyl cyclase alpha2 subunit
Gene Symbols GUCY1A2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GUCY1A2 (aa 128-177).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2977
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.