Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GUCY1A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115335 Supplier Invitrogen™ Supplier No. PA5115335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Guanylyl Cyclase alpha 2.

Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha and a beta subunit, catalyzes the conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide and nitrovasodilator drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUCY1A2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GUCY1A2
Gene Accession No. P33402, Q9WVI4
Gene Alias 6330407I18Rik; A230060L24Rik; GC-SA2; GCS-alpha-2; guanylate cyclase 1 soluble subunit alpha 2; guanylate cyclase 1, soluble, alpha 2; guanylate cyclase soluble subunit alpha-2; GUC1A2; GUCSA2; Gucy1a2; soluble guanylyl cyclase alpha2 subunit
Gene Symbols GUCY1A2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GUCY1A2(Accession P33402), corresponding to amino acid residues H118-E168.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234889, 2977, 66012
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.