Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GUCY1A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575446 Supplier Invitrogen™ Supplier No. PA575446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Guanylate cyclases belong to the adenylyl cyclase class-4/guanylyl cyclase family. There are two forms of guanylate cyclase, a soluble form (GCS or sGC), which act as receptors for nitric oxide and a membrane-bound receptor form (GC), which are peptide hormone receptors. The GC-C protein is composed of an extracellular domain, a single transmembrane domain, and a cytoplasmic region consisting of a kinase-like domain and a catalytic domain. It is expressed as two differentially glycosylated forms, a 130 kDa precursor form present in the endoplasmic reticulum and a 145 kDa form present on the plasma membrane. Ligand binding to the extracellular domain of GC-C promotes the accumulation of cGMP. GC-C acts as the receptor for heatstable enterotoxins, small peptides secreted by some pathogenic strains of E. coli that cause severe secretory diarrhea. GC-C also binds to guanylin and uroguanylin peptides, which modulate renal function in response to oral salt load.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUCY1A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GUCY1A1
Gene Accession No. P19686, Q02108, Q9ERL9
Gene Alias 1200016O07Rik; 4.6.1.2; alpha 1 sGC; GC-SA3; GCSalpha1; GC-S-alpha-1; GCS-alpha1; GCS-alpha-1; GCS-alpha-3; guanylate cyclase 1 soluble subunit alpha; guanylate cyclase 1, soluble, alpha 3; Guanylate cyclase soluble alpha 1 (GTP pyrophosphate - lyase); Guanylate cyclase soluble subunit alpha-1; Guanylate cyclase soluble subunit alpha-3; Guanylate cyclase, soluble, alpha 1 (GTP pyrophosphate - lyase); GUC1A3; GUCA3; GUCSA3; Gucy1a1; GUCY1A3; MYMY6; SGC; sGC-alpha1; soluble guanylate cyclase large subunit
Gene Symbols GUCY1A1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa 400-450 of Human GUCY1A3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2982, 497757, 60596
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.