Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCY1B3 (GCS-b-1) Monoclonal Antibody (GC11), Abbomax

Catalog No. 502204307
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4307 Supplier Abbomax Inc Supplier No. 604980
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Fetal lung tissue lysate Cellular location: Cytoplasm.

GUCY1B3 (GCS-b-1) Monoclonal antibody specifically detects GUCY1B3 (GCS-b-1) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen GUCY1B3 (GCS-b-1)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GC11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene GUCY1B1
Gene Accession No. Q02153
Gene Alias beta 1 sGC; GCbeta1; GCS beta1; GCS beta3; GC-SB3; GCSbeta1; GC-S-beta-1; GCS-beta1; GCS-beta-1; GCSbeta3; GCS-beta3; GCS-beta-3; guanylate cyclase 1 soluble subunit beta; guanylate cyclase 1 soluble subunit beta 1; guanylate cyclase 1, soluble, beta 3; guanylate cyclase 70 kDa subunit; guanylate cyclase soluble subunit beta-1; Guanylate cyclase soluble subunit beta-3; guanylate cyclase, soluble, beta 1; GUC1B3; GUCB3; GUCSB3; Gucy1b1; Gucy1b3; nitric oxide-sensitive guanylyl cyclase beta 1 subunit; NO-sensitive GC beta 1 subunit; NO-sensitive guanylyl cyclase beta 1 subunit; SGC; sgc1b1; si:ch73-34b5.3; soluble guanylate cyclase 1 beta 3; soluble guanylate cyclase beta-1 subunit; soluble guanylate cyclase small subunit; soluble guanylate cyclase small subunit beta 1; soluble guanylyl cyclase beta1 subunit
Host Species Mouse
Immunogen A synthetic peptide of C-terminus of human GUCY1B3
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.