Learn More
Invitrogen™ GUCY1B3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total Guanylyl Cyclase beta protein.
Specifications
Specifications
| Antigen | GUCY1B3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | GUCY1B1 |
| Gene Accession No. | O54865, P20595, Q02153 |
| Gene Alias | beta 1 sGC; GCbeta1; GCS beta1; GCS beta3; GC-SB3; GCSbeta1; GC-S-beta-1; GCS-beta1; GCS-beta-1; GCSbeta3; GCS-beta3; GCS-beta-3; guanylate cyclase 1 soluble subunit beta; guanylate cyclase 1 soluble subunit beta 1; guanylate cyclase 1, soluble, beta 3; guanylate cyclase 70 kDa subunit; guanylate cyclase soluble subunit beta-1; Guanylate cyclase soluble subunit beta-3; guanylate cyclase, soluble, beta 1; GUC1B3; GUCB3; GUCSB3; Gucy1b1; Gucy1b3; nitric oxide-sensitive guanylyl cyclase beta 1 subunit; NO-sensitive GC beta 1 subunit; NO-sensitive guanylyl cyclase beta 1 subunit; SGC; sgc1b1; si:ch73-34b5.3; soluble guanylate cyclase 1 beta 3; soluble guanylate cyclase beta-1 subunit; soluble guanylate cyclase small subunit; soluble guanylate cyclase small subunit beta 1; soluble guanylyl cyclase beta1 subunit |
| Gene Symbols | GUCY1B1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.