Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GUCY1B3 Recombinant Rabbit Monoclonal Antibody (5O8D9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538086
Description
Positive Samples: U-87MG, Mouse lung, Mouse brain, Mouse heart, Rat brain Immunogen sequence: MYGFVNHALE LLVIRNYGPE VWEDIKKEAQ LDEEGQFLVR IIYDDSKTYD LVAAASKVLN LNAGEILQMF GKMFFVFCQE SGYDTILRVL GSNVREFLQN.
Guanylate cyclases belong to the adenylyl cyclase class-4/guanylyl cyclase family. There are two forms of guanylate cyclase, a soluble form (GCS or sGC), which act as receptors for nitric oxide and a membrane-bound receptor form (GC), which are peptide hormone receptors. The GC-C protein is composed of an extracellular domain, a single transmembrane domain, and a cytoplasmic region consisting of a kinase-like domain and a catalytic domain. It is expressed as two differentially glycosylated forms, a 130 kDa precursor form present in the endoplasmic reticulum and a 145 kDa form present on the plasma membrane. Ligand binding to the extracellular domain of GC-C promotes the accumulation of cGMP. GC-C acts as the receptor for heatstable enterotoxins, small peptides secreted by some pathogenic strains of E. coli that cause severe secretory diarrhea. GC-C also binds to guanylin and uroguanylin peptides, which modulate renal function in response to oral salt load.
Specifications
GUCY1B3 | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O54865, P20595, Q02153 | |
GUCY1B1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Guanylyl Cyclase beta 1 (GUCY1B3) (Q02153). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
5O8D9 | |
Unconjugated | |
GUCY1B1 | |
beta 1 sGC; GCbeta1; GCS beta1; GCS beta3; GC-SB3; GCSbeta1; GC-S-beta-1; GCS-beta1; GCS-beta-1; GCSbeta3; GCS-beta3; GCS-beta-3; guanylate cyclase 1 soluble subunit beta; guanylate cyclase 1 soluble subunit beta 1; guanylate cyclase 1, soluble, beta 3; guanylate cyclase 70 kDa subunit; guanylate cyclase soluble subunit beta-1; Guanylate cyclase soluble subunit beta-3; guanylate cyclase, soluble, beta 1; GUC1B3; GUCB3; GUCSB3; Gucy1b1; Gucy1b3; nitric oxide-sensitive guanylyl cyclase beta 1 subunit; NO-sensitive GC beta 1 subunit; NO-sensitive guanylyl cyclase beta 1 subunit; SGC; sgc1b1; si:ch73-34b5.3; soluble guanylate cyclase 1 beta 3; soluble guanylate cyclase beta-1 subunit; soluble guanylate cyclase small subunit; soluble guanylate cyclase small subunit beta 1; soluble guanylyl cyclase beta1 subunit | |
Rabbit | |
Affinity chromatography | |
RUO | |
25202, 2983, 54195 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction