Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GUCY2C Antibody (2543C), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB2157C
Description
GUCY2C Monoclonal antibody specifically detects GUCY2C in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
GUCY2C | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
2543C | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
EC 4.6.1, GC-C, guanylate cyclase 2C (heat stable enterotoxin receptor), Guanylyl cyclase C, GUC2CEC 4.6.1.2, heat-stable enterotoxin receptor, hSTAR, Intestinal guanylate cyclase, STARSTA receptor | |
Human embryonic kidney cell HEK293-derived human Guanylyl Cyclase C/GUCY2C, Ser21-Gln430, Accession # P25092 | |
0.1 mL | |
Protein Kinase | |
2984 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction