Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GUCY2C Antibody (4G12), Novus Biologicals™

Mouse Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | GUCY2C |
---|---|
Clone | 4G12 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
GUCY2C Monoclonal antibody specifically detects GUCY2C in Human samples. It is validated for Flow Cytometry, ELISASpecifications
GUCY2C | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 4.6.1, GC-C, guanylate cyclase 2C (heat stable enterotoxin receptor), Guanylyl cyclase C, GUC2CEC 4.6.1.2, heat-stable enterotoxin receptor, hSTAR, Intestinal guanylate cyclase, STARSTA receptor | |
Recombinant Human GUCY2C protein [UniProt P25092] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4G12 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Protein Kinase | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
2984 | |
IgG2a | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title