Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GUCY2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558043 Supplier Invitrogen™ Supplier No. PA558043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVRGETYLKG RGNETTYWLT GMKDQKFNLP TPPTVENQQR LQAEFSDMIA NSLQKRQAAG IRSQKPRRVA SYKKGTLEYL QLNTTD Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.

Receptor for the E.coli heat-stable enterotoxin (E.coli enterotoxin markedly stimulates the accumulation of cGMP in mammalian cells expressing GC-C). Also activated by the endogenous peptide guanylin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUCY2C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GUCY2C
Gene Accession No. P25092
Gene Alias AI893437; DIAR6; Gcc; GC-C; guanylate cyclase 2C; guanylyl cyclase C; GUC2C; GUCY2C; heat stable enterotoxin receptor; heat-stable enterotoxin receptor; hSTAR; intestinal guanylate cyclase; MECIL; MUCIL; STA receptor; STAR
Gene Symbols GUCY2C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GUCY2C. Recombinant protein control fragment (Product #RP-96527).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.