Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GUCY2C Recombinant Human Monoclonal Antibody (2E5)
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549967
Description
GUCY2C Recombinant Monoclonal Antibody for Flow, ELISA
Receptor for the E.coli heat-stable enterotoxin (E.coli enterotoxin markedly stimulates the accumulation of cGMP in mammalian cells expressing GC-C). Also activated by the endogenous peptide guanylin.
Specifications
GUCY2C | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P25092 | |
GUCY2C | |
Recombinant Human GUCY2C protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Neutralization | |
2E5 | |
Unconjugated | |
GUCY2C | |
AI893437; DIAR6; Gcc; GC-C; guanylate cyclase 2C; guanylyl cyclase C; GUC2C; GUCY2C; heat stable enterotoxin receptor; heat-stable enterotoxin receptor; hSTAR; intestinal guanylate cyclase; MECIL; MUCIL; STA receptor; STAR | |
Human | |
Affinity chromatography | |
RUO | |
2984 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction