Learn More
Invitrogen™ GUCY2D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109800
Description
Immunogen sequence: AVFTVGVLGP WACDPIFSRA RPDLAARLAA ARLNRDPGLA GGPRFEVALL PEPCRTPGSL GAVSSALARV SGLVGPVNPA ACRPAELLAE EAGIALVPWG CPWTQAEGTT APAVTPAADA LYALLRAFGW ARVALVTAPQ DLWVEAGRSL STALRARGLP VASVTSMEPL DLSGAREALR KVRDGPRVTA VIMVMHSVLL GGEEQRYLLE AAEELGLTDG SLVFLPFDTI HYALSPGPEA LAALANSSQL RRAHDAVLTL TRHCPSEGSV LDSLRRAQER RELPSDLNLQ QVSPLFGTI.
Probably plays a specific functional role in the rods and/or cones of photoreceptors. It may be the enzyme involved in the resynthesis of cGMP required for recovery of the dark state after phototransduction.
Specifications
GUCY2D | |
Polyclonal | |
Unconjugated | |
GUCY2D | |
cone rod dystrophy 6; CORD5; CORD6; CYGD; guanylate cyclase 2d; guanylate cyclase 2D, membrane (retina-specific); guanylate cyclase 2D, retinal; Guanylate cyclase, olfactory; guanylyl cyclase 2d; guanylyl cyclase D; GUC1A4; GUC2D; GUCY2D; LCA; LCA1; olfactory guanylyl cyclase GC-D; RCD2; RETGC; RETGC1; RETGC-1; retinal guanylate cyclase 1; retinal guanylyl cyclase 1; Rod outer segment membrane guanylate cyclase; ROSGC; ROS-GC; ROS-GC1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
113911, 14918, 3000 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51839, Q02846 | |
GUCY2D | |
Recombinant fusion protein containing a sequence corresponding to amino acids 52-350 of human GUCY2D (NP_000171.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.