Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GUF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552028 Supplier Invitrogen™ Supplier No. PA552028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VENQIEKVFD IPSDECIKIS AKLGTNVESV LQAIIERIPP PKVHRKNPLR ALVFDSTFDQ YRGVIANVAL FDGVVSKGDK IVSAHTQKTY EVNEVGVLNP NEQPTHKLYA GQVGYLIAGM KDVTEAQIGD TLCLHKQPVE PL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

Promotes mitochondrial protein synthesis. May act as a fidelity factor of the translation reaction, by catalyzing a one- codon backward translocation of tRNAs on improperly translocated ribosomes. Binds to mitochondrial ribosomes in a GTP-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GUF1
Gene Accession No. Q8N442
Gene Alias 4631409J12; AA407526; EF-4; elongation factor 4 homolog; fc88c11; GTP binding elongation factor GUF1; GTPase GUF1; GTPase of unknown function 1; GTP-binding protein GUF1 homolog; GTP-binding protein GUF1-like protein-like protein; Guf1; GUF1 GTPase homolog; GUF1 GTPase homolog (S. cerevisiae); GUF1 homolog, GTPase; GUF1 homolog, GTPase; translation factor GUF1, mitochondrial; im:6906935; RGD1310774; Ribosomal back-translocase; translation factor GUF1, mitochondrial; wu:fc88c11
Gene Symbols GUF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GUF1. Recombinant protein control fragment (Product #RP-102347).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60558
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.