Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GUF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570331 Supplier Invitrogen™ Supplier No. PA570331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Yeast: 90%; Zebrafish: 79%.

Promotes mitochondrial protein synthesis. May act as a fidelity factor of the translation reaction, by catalyzing a one- codon backward translocation of tRNAs on improperly translocated ribosomes. Binds to mitochondrial ribosomes in a GTP-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GUF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GUF1
Gene Accession No. Q8C3X4
Gene Alias 4631409J12; AA407526; EF-4; elongation factor 4 homolog; fc88c11; GTP binding elongation factor GUF1; GTPase GUF1; GTPase of unknown function 1; GTP-binding protein GUF1 homolog; GTP-binding protein GUF1-like protein-like protein; Guf1; GUF1 GTPase homolog; GUF1 GTPase homolog (S. cerevisiae); GUF1 homolog, GTPase; GUF1 homolog, GTPase; translation factor GUF1, mitochondrial; im:6906935; RGD1310774; Ribosomal back-translocase; translation factor GUF1, mitochondrial; wu:fc88c11
Gene Symbols GUF1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse GUF1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231279
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.