Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELN, Guinea Pig, Polyclonal Antibody, Abnova™

Catalog No. 89116716 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-716 Supplier Abnova Corporation Supplier No. PAB13514
Add to Cart
Add to Cart

Guinea pig polyclonal antibody raised against native ELN.

This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen ELN
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Guinea pig polyclonal antibody raised against native ELN.
Dilution ELISA (>1:200) Immunohistochemistry (1:500-1000) Immunofluorescence (1:80) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from phosphate buffered solution
Gene ELN
Gene Alias FLJ38671/FLJ43523/SVAS/WBS/WS
Gene Symbols ELN
Host Species Guinea Pig
Immunogen Native purified ELN from human skin.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2006
Test Specificity This antibody reacts with human elastin 100%, cross-reactivity in RIA at 1:500 dilution: human collagen type I, II, III, IV, V <0.1 %; human fibronectin <0.1%.
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.