Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guinea Pig anti-Chlamydia (Genus Specific), Polyclonal, Abnova™

Catalog No. 89137730 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-730 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-730 Supplier Abnova Corporation Supplier No. PAB13908

Specifications

Antigen Chlamydia (Genus Specific)
Applications Immunoprecipitation, Immunoblot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation In antiserum
Host Species Guinea Pig
Immunogen Elementary bodies from LGV2 serovar.
Quantity 1 mL
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.