Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium (Genus Specific), Guinea pig, Polyclonal Antibody, Abnova™

Catalog No. 89137796 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-796 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-796 Supplier Abnova Corporation Supplier No. PAB14008

Guinea pig polyclonal antibody raised against Mycobacterium (Genus Specific).

Specifications

Antigen Mycobacterium (Genus Specific)
Applications ELISA, Immunoblot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Guinea pig polyclonal antibody raised against Mycobacterium (Genus Specific).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Guinea Pig
Immunogen Native genus specific antigens from Mycobacterium tuberculosis.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts primarily against lipo-arabinomannan (LAM). Reacts with several membrane proteins by western blot. Cross-reacts against all Mycobacterial species.
Target Species Bacteria
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.