Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LHB, Guinea pig, Polyclonal Antibody, Abnova™

Catalog No. 89116993 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-993 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-993 Supplier Abnova Corporation Supplier No. PAB14309
Add to Cart
Add to Cart

Guinea pig polyclonal antibody raised against synthetic peptide of LHB.

Specifications

Antigen LHB
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Description Guinea pig polyclonal antibody raised against synthetic peptide of LHB.
Dilution Immunohistochemistry (1:3000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene LHB
Gene Symbols LHB
Host Species Guinea Pig
Immunogen A synthetic peptide corresponding to sheep LHB.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 443395
Target Species Sheep
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.