Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXT, Guinea pig, Polyclonal Antibody, Abnova™

Catalog No. 89116994 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-994 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-994 Supplier Abnova Corporation Supplier No. PAB14310
Add to Cart
Add to Cart

Guinea pig polyclonal antibody raised against synthetic peptide of OXT.

There are two proteins encoded by this gene, oxytocin and neurophysin I. Oxytocin is posterior pituitary hormone which is synthesized as an inactive precursor in the hypothalamus along with its carrier protein neurophysin I. Together with neurophysin, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis, where it is either stored or secreted into the bloodstream. The precursor seems to be activated while it is being transported along the axon to the posterior pituitary. This hormone contracts smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions. [provided by RefSeq

Sequence: YIQNCPLG

Specifications

Antigen OXT
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Description Guinea pig polyclonal antibody raised against synthetic peptide of OXT.
Dilution Immunohistochemistry (1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene OXT
Gene Alias MGC126890/MGC126892/OT/OT-NPI
Gene Symbols OXT
Host Species Guinea Pig
Immunogen A synthetic peptide (conjugated with carrier protein) corresponding to amino acids 20-28 of human OXT.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5020
Target Species Human, Sheep
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.