Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYY, Guinea pig, Polyclonal Antibody, Abnova™

Catalog No. 89116846 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-846 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-846 Supplier Abnova Corporation Supplier No. PAB13801
Add to Cart
Add to Cart

Guinea pig polyclonal antibody raised against synthetic peptide of PYY.

Specifications

Antigen PYY
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Guinea pig polyclonal antibody raised against synthetic peptide of PYY.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1500-1:2500) Immunohistochemistry (Frozen sections) (1:1500-1:2500) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from antiserum
Gene PYY
Gene Symbols PYY
Host Species Guinea Pig
Immunogen A synthetic peptide corresponding to pig PYY.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445018
Target Species Feline, Mouse, Pig, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.